missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Bromo-2-fluorophenyl)-2-methoxybenzamide, 97%

Brand:  Alfa Aesar™ H59930.03

Code : NEW

Additional Details : CAS Number : 420092-67-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416388

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 30-07-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Bromo-2-fluorophenyl)-2-methoxybenzamide
420092-67-7
MFCD01343370

Specifications

N-(4-Bromo-2-fluorophenyl)-2-methoxybenzamide
420092-67-7
97%
MFCD01343370
YSGDYHBVXLMUHJ-UHFFFAOYSA-N
324.149
324.15
Solid
C14H11BrFNO2
1g
COC1=CC=CC=C1C(=O)NC2=C(C=C(C=C2)Br)F
868874
Videos
SDS