missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Ethylphenyl)-3,5-dinitrobenzamide, 97%

Brand:  Alfa Aesar™ H59677.03

Code : NEW

Additional Details : CAS Number : 36293-11-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15476058

  • 646.00 € / 1g

Estimated Shipment date
from Supplier 17-09-2019

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

36293-11-5
315.285
KNQZDRMQDPWSQK-UHFFFAOYSA-N
2057610
CCC1=CC=C(C=C1)NC(=O)C2=CC(=CC(=C2)[N+](=O)[O-])[N+](=O)[O-]
C15H13N3O5
MFCD00783642
n-4-ethylphenyl-3,5-dinitrobenzamide, 3,5-dinitrophenyl-n-4-ethylphenyl carboxamide
N-(4-ethylphenyl)-3,5-dinitrobenzamide

Specifications

N-(4-Ethylphenyl)-3,5-dinitrobenzamide
1g
C15H13N3O5
2896164
KNQZDRMQDPWSQK-UHFFFAOYSA-N
N-(4-ethylphenyl)-3,5-dinitrobenzamide
2057610
Solid
Brown
36293-11-5
MFCD00783642
n-4-ethylphenyl-3,5-dinitrobenzamide, 3,5-dinitrophenyl-n-4-ethylphenyl carboxamide
CCC1=CC=C(C=C1)NC(=O)C2=CC(=CC(=C2)[N+](=O)[O-])[N+](=O)[O-]
315.285
315.28
97%
Videos
SDS Documents