missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Fluorophenyl)-3,5-dinitrobenzamide, 97%

Brand:  Alfa Aesar™ H59754.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416158

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Fluorophenyl)-3,5-dinitrobenzamide
305.22
MFCD00587406

Specifications

N-(4-Fluorophenyl)-3,5-dinitrobenzamide
97%
MFCD00587406
Solid
305.22
1g
Videos
SDS