missing translation for 'onlineSavingsMsg'
Learn More

N-(5-chloro-2-pyridyl)bis(trifluoromethanesulfonimide), Thermo Scientific Chemicals

Catalog Number p-3593471
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
145100-51-2
C7H3ClF6N2O4S2
392.69
MFCD00191833
TUFGVZMNGTYAQD-UHFFFAOYSA-N
2-n,n-bis trifluoromethylsulphonyl amino-5-chloropyridine, 2-n,n-bis trifluoromethanesulfonyl amino-5-chloropyridine, comins' reagent, n-5-chloro-2-pyridyl triflimide, 2-n,n-bis trifluoromethylsulfonyl amino-5-chloropyridine, comins triflating reagent, n-5-chloro-2-pyridyl bis trifluoromethanesulfonimide, n-5-chloropyridin-2-yl-1,1,1-trifluoro-n-trifluoromethylsulfonyl methanesulfonamide, n-5-chloropyridin-2-yl triflimide, n-5-chloropyridin-2-yl-1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide
388544
N-(5-chloropyridin-2-yl)-1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
C1=CC(=NC=C1Cl)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F
This item is not returnable. View return policy
145100-51-2
C7H3ClF6N2O4S2
392.69
MFCD00191833
TUFGVZMNGTYAQD-UHFFFAOYSA-N
2-n,n-bis trifluoromethylsulphonyl amino-5-chloropyridine, 2-n,n-bis trifluoromethanesulfonyl amino-5-chloropyridine, comins' reagent, n-5-chloro-2-pyridyl triflimide, 2-n,n-bis trifluoromethylsulfonyl amino-5-chloropyridine, comins triflating reagent, n-5-chloro-2-pyridyl bis trifluoromethanesulfonimide, n-5-chloropyridin-2-yl-1,1,1-trifluoro-n-trifluoromethylsulfonyl methanesulfonamide, n-5-chloropyridin-2-yl triflimide, n-5-chloropyridin-2-yl-1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide
388544
N-(5-chloropyridin-2-yl)-1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
C1=CC(=NC=C1Cl)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY
Melting Point 45.0°C to 47.0°C
Boiling Point 77.0°C to 90.0°C (0.2 mmHg)
Packaging Glass bottle
Quantity 1 g
Formula Weight 392.69
Chemical Name or Material N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulfonimide)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'