missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-DL-tryptophan, 98+%, extra pure

Catalog Number p-50009
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
87-32-1
C13H14N2O3
246.27
MFCD00005644
DZTHIGRZJZPRDV-UHFFFAOYSA-N
n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl
2002
CHEBI:70976
2-acetamido-3-(1H-indol-3-yl)propanoic acid
CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
87-32-1
C13H14N2O3
246.27
MFCD00005644
DZTHIGRZJZPRDV-UHFFFAOYSA-N
n-acetyl-dl-tryptophan, n-acetyltryptophan, ac-dl-trp-oh, acetyltryptophan, dl-acetyltryptophan, 2-acetamido-3-1h-indol-3-yl propanoic acid, dl-n-acetyltryptophan, n-acetyl-dl-tryptophane, l-tryptophan, n-acetyl, dl-tryptophan, n-acetyl
2002
CHEBI:70976
2-acetamido-3-(1H-indol-3-yl)propanoic acid
CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 204.0°C to 206.0°C
Color White to Yellow
Loss on Drying 0.5% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: 0,51g/L (20°C). Other solubilities: soluble in methanol
Formula Weight 246.27
Percent Purity 98+%
Grade Extra Pure
Physical Form Fine Crystalline Powder
Chemical Name or Material N-Acetyl-DL-tryptophan, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'