missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-glutamine, 97%, Acros Organics

Brand:  Acros Organics 451641000

Code : NEW

Additional Details : CAS Number : 35305-74-9 Weight : 0.10000kg

Packaging Glass bottle
Quantity 100g

 View more versions of this product

Product Code. 15264884

  • 261.30 € / 100g

Estimated Shipment
20-07-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

35305-74-9
188.18
n-acetyl-l-glutamine, aceglutamide, l-glutamine, n-acetyl, nalpha-acetyl-l-glutamine, s-2-acetamido-5-amino-5-oxopentanoic acid, acetylglutamine, n-alpha-acetyl-l-glutamine, n-acetylglutamine, acutil-s, acetyl-l-glutamine
CHEBI:21553
CC(=O)NC(CCC(=O)N)C(=O)O
C7H12N2O4
KSMRODHGGIIXDV-YFKPBYRVSA-N
182230
(2S)-2-acetamido-5-amino-5-oxopentanoic acid

Specifications

N-Acetyl-L-glutamine
0.2% max
100g
97%
n-acetyl-l-glutamine, aceglutamide, l-glutamine, n-acetyl, nalpha-acetyl-l-glutamine, s-2-acetamido-5-amino-5-oxopentanoic acid, acetylglutamine, n-alpha-acetyl-l-glutamine, n-acetylglutamine, acutil-s, acetyl-l-glutamine
KSMRODHGGIIXDV-YFKPBYRVSA-N
(2S)-2-acetamido-5-amino-5-oxopentanoic acid
182230
188.18
Glass bottle
206°C
35305-74-9
C7H12N2O4
Solubility in water: soluble
CC(=O)NC(CCC(=O)N)C(=O)O
188.18
CHEBI:21553
97%
Videos
Documents