missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-Boc-D-glutamine, 98+%, Thermo Scientific Chemicals

Product Code. p-7044866
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11388215

Brand: Thermo Scientific Alfa Aesar L08536.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

N2-Boc-D-glutamine is an N-Boc-protected form of D-Glutamine. D-Glutamine is an unnatural isomer of L-Glutamine that is present in human plasma an is a source of liberated ammonia. D-Glutamine can be synthesized by enzymatic means or can be found in cheeses, wine and vinegars as well.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N2-Boc-D-glutamine is an N-Boc-protected form of D-Glutamine. D-Glutamine is an unnatural isomer of L-Glutamine that is present in human plasma an is a source of liberated ammonia. D-Glutamine can be synthesized by enzymatic means or can be found in cheeses, wine and vinegars as well.

Solubility
Soluble, 1mmol in 2mL DMF. Soluble in chloroform and methanol.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61348-28-5
Molecular Formula C10H18N2O5
Molecular Weight (g/mol) 246.263
MDL Number MFCD00038158
InChI Key VVNYDCGZZSTUBC-ZCFIWIBFSA-N
Synonym boc-d-glutamine, boc-d-gln-oh, boc-d-gln, n alpha-boc-d-glutamine, n-alpha-t-butoxycarbonyl-d-glutamine, n-tert-butoxycarbonyl-d-glutamine, r-5-amino-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-l-gln-oh, n-alpha-t-butyloxycarbonyl-d-glutamine, n-alpha-tert-butyloxycarbonyl-d-glutamine
PubChem CID 2724760
IUPAC Name (2R)-5-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC(=O)N)C(=O)O

Specifications

Melting Point 117°C to 119°C
Beilstein 1981311
Solubility Information Soluble,1mmol in 2mL DMF. Soluble in chloroform and methanol.
Quantity 250 mg
Formula Weight 246.26
Optical Rotation +3.5° (c=2 in Ethanol)
Percent Purity ≥98%
Chemical Name or Material N(alpha)-Boc-D-glutamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.