missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Benzyl-2-fluoro-N-isopropylbenzamide, 97%

Brand:  Alfa Aesar™ H59672.03

Code : NEW

Additional Details : CAS Number : 312924-64-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416058

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 21-11-2018
Add to basket

Specifications

Chemical Identifiers

312924-64-4
271.335
XZFUSRGNLTUTPS-UHFFFAOYSA-N
579323
CC(C)N(CC1=CC=CC=C1)C(=O)C2=CC=CC=C2F
C17H18FNO
MFCD01214067
n-benzyl-2-fluoro-n-isopropylbenzamide, n-benzyl-2-fluoro-n-isopropylbenzamide #, benzamide, n-benzyl-2-fluoro-n-isopropyl, n-benzyl-2-fluoro-n-propan-2-yl benzamide, 2-fluorophenyl-n-methylethyl-n-benzylcarboxamide
N-benzyl-2-fluoro-N-propan-2-ylbenzamide

Specifications

N-Benzyl-2-fluoro-N-isopropylbenzamide
1g
C17H18FNO
n-benzyl-2-fluoro-n-isopropylbenzamide, n-benzyl-2-fluoro-n-isopropylbenzamide #, benzamide, n-benzyl-2-fluoro-n-isopropyl, n-benzyl-2-fluoro-n-propan-2-yl benzamide, 2-fluorophenyl-n-methylethyl-n-benzylcarboxamide
CC(C)N(CC1=CC=CC=C1)C(=O)C2=CC=CC=C2F
271.335
271.33
97%
Yellow
312924-64-4
MFCD01214067
XZFUSRGNLTUTPS-UHFFFAOYSA-N
N-benzyl-2-fluoro-N-propan-2-ylbenzamide
579323
Liquid
Videos
SDS