missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-L-leucine, 98% (dry wt.), may cont. up to ca 5% solvent, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
It is employed as a reactant for peptide-based preparation of potential antitumor agents, preparation of cyclopropyl peptidomimetics and peptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a reactant for peptide-based preparation of potential antitumor agents, preparation of cyclopropyl peptidomimetics and peptide synthesis.
Solubility
Not miscible or difficult to mix in water. Soluble in chloroform, DMSO and methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Keep it refrigerated.
It is employed as a reactant for peptide-based preparation of potential antitumor agents, preparation of cyclopropyl peptidomimetics and peptide synthesis.
Solubility
Not miscible or difficult to mix in water. Soluble in chloroform, DMSO and methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Keep it refrigerated.
Specifications
Density | 1 |
Flash Point | >110°C (230°F) |
Odor | Odorless |
Refractive Index | 1.508 |
Quantity | 5 g |
Beilstein | 1253861 |
Solubility Information | Not miscible or difficult to mix in water. Soluble in chloroform,DMSO and methanol. |
Optical Rotation | −17.5° (c=2 in Ethanol) |
Formula Weight | 265.3 |
Percent Purity | ≥98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'