missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1,6-diaminohexane, 98%, Thermo Scientific Chemicals

Catalog Number p-7047909
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
51857-17-1
C11H24N2O2
216.325
MFCD00671489
RVZPDKXEHIRFPM-UHFFFAOYSA-N
n-boc-1,6-diaminohexane, n-boc-1,6-hexanediamine, tert-butyl n-6-aminohexyl carbamate, tert-butyl 6-aminohexyl carbamate, n-tert-butoxycarbonyl-1,6-diaminohexane, n-tert-butoxycarbonyl-1,6-hexanediamine, tert-butyl 6-aminohexylcarbamate, boc-1,6-diaminohexane hydrochloride, 6-tert-butoxycarbonylamino hexylamine, n-6-aminohexyl carbamic acid tert-butyl ester
2733170
tert-butyl N-(6-aminohexyl)carbamate
CC(C)(C)OC(=O)NCCCCCCN
This item is not returnable. View return policy
51857-17-1
C11H24N2O2
216.325
MFCD00671489
RVZPDKXEHIRFPM-UHFFFAOYSA-N
n-boc-1,6-diaminohexane, n-boc-1,6-hexanediamine, tert-butyl n-6-aminohexyl carbamate, tert-butyl 6-aminohexyl carbamate, n-tert-butoxycarbonyl-1,6-diaminohexane, n-tert-butoxycarbonyl-1,6-hexanediamine, tert-butyl 6-aminohexylcarbamate, boc-1,6-diaminohexane hydrochloride, 6-tert-butoxycarbonylamino hexylamine, n-6-aminohexyl carbamic acid tert-butyl ester
2733170
tert-butyl N-(6-aminohexyl)carbamate
CC(C)(C)OC(=O)NCCCCCCN

N-Boc-1,6-diaminohexane is used to prepare 1,3-Bis[6-(Boc-amino)hexyl]urea by reacting with carbonyl dichloride in the presence of triethylamine. Further, it is used as a reagent for the introduction of a C6-spacer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-1,6-diaminohexane is used to prepare 1,3-Bis[6-(Boc-amino)hexyl]urea by reacting with carbonyl dichloride in the presence of triethylamine. Further, it is used as a reagent for the introduction of a C6-spacer.

Solubility
Soluble in dichloromethane and ethyl acetate.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Boiling Point 106°C to 110°C (0.3mmHg)
UN Number UN2735
Beilstein 2089264
Refractive Index 1.461
Solubility Information Soluble in dichloromethane and ethyl acetate.
Formula Weight 216.33
Percent Purity 98%
Density 0.965
Quantity 1 g
Flash Point >110°C (230°F)
Sensitivity Air Sensitive
Chemical Name or Material N-Boc-1,6-diaminohexane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'