missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-methyl-D-phenylalanine, 95%, Thermo Scientific™

Catalog Number p-7041569
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
114873-14-2
C15H21NO4
279.336
MFCD01862943
HBBXWMALJNZDOM-GFCCVEGCSA-N
boc-3-methyl-d-phenylalanine, boc-d-phe 3-me-oh, boc-d-3-methylphenylalanine, r-2-tert-butoxycarbonyl amino-3-m-tolyl propanoic acid, boc-d-3-methylphe, boc-3-methy-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-3-methylphenyl propanoic acid, boc-m-me-d-phe-oh, boc-d-3-me-phe-oh, boc-d-3-methyl phenylalanine
7010034
(2R)-3-(3-methylphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC1=CC(=CC=C1)CC(C(=O)O)NC(=O)OC(C)(C)C
This item is not returnable. View return policy
114873-14-2
C15H21NO4
279.336
MFCD01862943
HBBXWMALJNZDOM-GFCCVEGCSA-N
boc-3-methyl-d-phenylalanine, boc-d-phe 3-me-oh, boc-d-3-methylphenylalanine, r-2-tert-butoxycarbonyl amino-3-m-tolyl propanoic acid, boc-d-3-methylphe, boc-3-methy-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-3-methylphenyl propanoic acid, boc-m-me-d-phe-oh, boc-d-3-me-phe-oh, boc-d-3-methyl phenylalanine
7010034
(2R)-3-(3-methylphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC1=CC(=CC=C1)CC(C(=O)O)NC(=O)OC(C)(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 279.33
Percent Purity 95%
Chemical Name or Material N-Boc-3-methyl-D-phenylalanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'