missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-Allylamine, 98%, Thermo Scientific Chemicals

Catalog Number 10668540
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
78888-18-3
C8H15NO2
157.21
AWARHXCROCWEAK-UHFFFAOYSA-N
tert-butyl allylcarbamate, tert-butyl n-allylcarbamate, tert-butyl n-prop-2-en-1-yl carbamate, n-boc-allylamine, t-butyl n-allylcarbamate, carbamic acid, 2-propenyl-, 1,1-dimethylethyl ester, acmc-209pfu, allyl tert-butyl carbamate, tert-butyl-n-allylcarbamate, n-tertbutoxycarbonyl allyl amine
4067274
tert-butyl N-prop-2-enylcarbamate
CC(C)(C)OC(=O)NCC=C
This item is not returnable. View return policy
78888-18-3
C8H15NO2
157.21
AWARHXCROCWEAK-UHFFFAOYSA-N
tert-butyl allylcarbamate, tert-butyl n-allylcarbamate, tert-butyl n-prop-2-en-1-yl carbamate, n-boc-allylamine, t-butyl n-allylcarbamate, carbamic acid, 2-propenyl-, 1,1-dimethylethyl ester, acmc-209pfu, allyl tert-butyl carbamate, tert-butyl-n-allylcarbamate, n-tertbutoxycarbonyl allyl amine
4067274
tert-butyl N-prop-2-enylcarbamate
CC(C)(C)OC(=O)NCC=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 34.9°C to 38.3°C
Boiling Point 48.0°C to 50.0°C (1.0mmHg)
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Assay Percent Range 97.5% min. (GC)
Infrared Spectrum Authentic
Packaging Glass Bottle
Formula Weight 157.21
Percent Purity 98%
Quantity 10 g
Flash Point 79°C
Physical Form Low Melting Solid
Chemical Name or Material N-BOC-Allylamine
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-allylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'