missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-Hexahydro-1H-azepin-4-one, 97%, ACROS Organics™

73.10 € - 251.10 €

Chemical Identifiers

CAS 188975-88-4
Molecular Formula C11H19NO3
Molecular Weight (g/mol) 213.28
InChI Key PMLBUVZPRKXMOX-UHFFFAOYSA-N
Synonym n-boc-hexahydro-1h-azepin-4-one, 1-boc-hexahydro-1h-azepin-4-one, 4-oxoazepane-1-carboxylic acid tert-butyl ester, 1-boc-homopiperazin-4-one, 1-boc-4-azepanone, n-boc-hexahydro-1h-azepine-4-one, tert-butyl hexahydro-4-oxo-1h-azepine-1-carboxylate, 1-tert-butoxycarbonyl-4-oxoazepane, 4-oxo-azepane-1-carboxylic acid tert-butyl ester, 1,1-dimethylethyl 4-oxohexahydro-1h-azepine-1-carboxylate
PubChem CID 1512679
IUPAC Name tert-butyl 4-oxoazepane-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(=O)CC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10311200 SDS View Documents Acros Organics
435540010
1g Glass bottle 73.10 €
1g View alternative products
Save Up to 
»
Estimated Shipment
27-07-2020
Please to purchase this item. Need a web account? Register with us today!
10008339 SDS View Documents Acros Organics
435540050
5g Glass bottle 251.10 €
5g View alternative products
Save Up to 
»
Estimated Shipment
27-07-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

188975-88-4
213.28
n-boc-hexahydro-1h-azepin-4-one, 1-boc-hexahydro-1h-azepin-4-one, 4-oxoazepane-1-carboxylic acid tert-butyl ester, 1-boc-homopiperazin-4-one, 1-boc-4-azepanone, n-boc-hexahydro-1h-azepine-4-one, tert-butyl hexahydro-4-oxo-1h-azepine-1-carboxylate, 1-tert-butoxycarbonyl-4-oxoazepane, 4-oxo-azepane-1-carboxylic acid tert-butyl ester, 1,1-dimethylethyl 4-oxohexahydro-1h-azepine-1-carboxylate
tert-butyl 4-oxoazepane-1-carboxylate
C11H19NO3
PMLBUVZPRKXMOX-UHFFFAOYSA-N
1512679
CC(C)(C)OC(=O)N1CCCC(=O)CC1

Specifications

N-BOC-Hexahydro-1H-azepin-4-one
Glass bottle
1g
96.0
96% min. (GC)
n-boc-hexahydro-1h-azepin-4-one, 1-boc-hexahydro-1h-azepin-4-one, 4-oxoazepane-1-carboxylic acid tert-butyl ester, 1-boc-homopiperazin-4-one, 1-boc-4-azepanone, n-boc-hexahydro-1h-azepine-4-one, tert-butyl hexahydro-4-oxo-1h-azepine-1-carboxylate, 1-tert-butoxycarbonyl-4-oxoazepane, 4-oxo-azepane-1-carboxylic acid tert-butyl ester, 1,1-dimethylethyl 4-oxohexahydro-1h-azepine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(=O)CC1
213.28
213.28
97%
Authentic
Colorless to Yellow
188975-88-4
100.0
C11H19NO3
PMLBUVZPRKXMOX-UHFFFAOYSA-N
tert-butyl 4-oxoazepane-1-carboxylate
1512679
Liquid or Low Melting Solid
Safety and Handling

Safety and Handling

Product Identifier
  • N-BOC-Hexahydro-1H-azepin-4-one

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.