missing translation for 'onlineSavingsMsg'
Learn More

N-α-BOC-L-Asparagine 4-nitrophenyl ester, 98%, ACROS Organics™

Brand:  Acros Organics 440850050

Code : 29

Additional Details : CAS Number : 4587-33-1 Weight : 0.05500kg

Quantity 5g

 View more versions of this product

Product Code. 11935511

  • 44.40 € / 5g

Estimated Shipment: 13-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

4587-33-1
353.33
boc-asn-onp, boc-l-asparagine 4-nitrophenyl ester, boc-asn-ophno2, s-4-nitrophenyl 4-amino-2-tert-butoxycarbonyl amino-4-oxobutanoate, 4-nitrophenyl 2s-2-tert-butoxycarbonyl amino-3-carbamoylpropanoate, pubchem12095, threonin-tert.-butylether, ch3 3coconhch cooh ch2cooh, n1-t-boc c1-p-nitrophenol asparagine, boc-asn-onp hplc
CHEBI:3147
CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)OC1=CC=C(C=C1)[N+](=O)[O-]
C15H19N3O7
IAPXDJMULQXGDD-NSHDSACASA-N
439663
(4-nitrophenyl) (2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoate

Specifications

N-α-BOC-L-Asparagine 4-nitrophenyl ester
-36.4
White
5g
97.5
97.5% min. (HPLC)
boc-asn-onp, boc-l-asparagine 4-nitrophenyl ester, boc-asn-ophno2, s-4-nitrophenyl 4-amino-2-tert-butoxycarbonyl amino-4-oxobutanoate, 4-nitrophenyl 2s-2-tert-butoxycarbonyl amino-3-carbamoylpropanoate, pubchem12095, threonin-tert.-butylether, ch3 3coconhch cooh ch2cooh, n1-t-boc c1-p-nitrophenol asparagine, boc-asn-onp hplc
CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)OC1=CC=C(C=C1)[N+](=O)[O-]
353.33
CHEBI:3147
Powder
Authentic
1% Max.
136°C to 140°C
4587-33-1
100.0
C15H19N3O7
IAPXDJMULQXGDD-NSHDSACASA-N
(4-nitrophenyl) (2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoate
439663
353.33
98%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • N-alpha-BOC-L-Asparagine 4-nitrophenyl ester

SDS Documents
Certificates