missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-N-methyl-1,3-diaminopropane, 95%

Brand:  Alfa Aesar™ H66932.MD

Code : NEW

Additional Details : CAS Number : 150349-36-3 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15472839

  • 49.30 € / 250mg

Estimated Shipment date
from Supplier 17-09-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

150349-36-3
188.271
PNQYAMWGTGWJDW-UHFFFAOYSA-N
2756543
CC(C)(C)OC(=O)N(C)CCCN
C9H20N2O2
MFCD01631189
n-3-aminopropyl-n-methylcarbamic acid tert-butyl ester, tert-butyl n-3-aminopropyl-n-methylcarbamate, tert-butyl 3-aminopropyl methyl carbamate, 3-n-boc-n-methylamino propylamine, n-boc-n-methyl-1,3-diaminopropane, n-3-amino-propyl-n-methylcarbamic acid t-butyl ester, acid tert-butyl ester, 3-n-tert-butoxycarbonyl-n-methylamino propylamine, tert-butyl n-3-aminopropyl-n-methyl carbamate
tert-butyl N-(3-aminopropyl)-N-methylcarbamate

Specifications

N-Boc-N-methyl-1,3-diaminopropane
250mg
C9H20N2O2
n-3-aminopropyl-n-methylcarbamic acid tert-butyl ester, tert-butyl n-3-aminopropyl-n-methylcarbamate, tert-butyl 3-aminopropyl methyl carbamate, 3-n-boc-n-methylamino propylamine, n-boc-n-methyl-1,3-diaminopropane, n-3-amino-propyl-n-methylcarbamic acid t-butyl ester, acid tert-butyl ester, 3-n-tert-butoxycarbonyl-n-methylamino propylamine, tert-butyl n-3-aminopropyl-n-methyl carbamate
CC(C)(C)OC(=O)N(C)CCCN
188.271
188.27
95%
Yellow
150349-36-3
MFCD01631189
PNQYAMWGTGWJDW-UHFFFAOYSA-N
tert-butyl N-(3-aminopropyl)-N-methylcarbamate
2756543
Liquid
Videos
SDS Documents