missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-N-methyl-1,3-diaminopropane, 95%, Thermo Scientific Chemicals

Catalog Number p-7057348
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
150349-36-3
C9H20N2O2
188.271
MFCD01631189
PNQYAMWGTGWJDW-UHFFFAOYSA-N
n-3-aminopropyl-n-methylcarbamic acid tert-butyl ester, tert-butyl n-3-aminopropyl-n-methylcarbamate, tert-butyl 3-aminopropyl methyl carbamate, 3-n-boc-n-methylamino propylamine, n-boc-n-methyl-1,3-diaminopropane, n-3-amino-propyl-n-methylcarbamic acid t-butyl ester, acid tert-butyl ester, 3-n-tert-butoxycarbonyl-n-methylamino propylamine, tert-butyl n-3-aminopropyl-n-methyl carbamate
2756543
tert-butyl N-(3-aminopropyl)-N-methylcarbamate
CC(C)(C)OC(=O)N(C)CCCN
This item is not returnable. View return policy
150349-36-3
C9H20N2O2
188.271
MFCD01631189
PNQYAMWGTGWJDW-UHFFFAOYSA-N
n-3-aminopropyl-n-methylcarbamic acid tert-butyl ester, tert-butyl n-3-aminopropyl-n-methylcarbamate, tert-butyl 3-aminopropyl methyl carbamate, 3-n-boc-n-methylamino propylamine, n-boc-n-methyl-1,3-diaminopropane, n-3-amino-propyl-n-methylcarbamic acid t-butyl ester, acid tert-butyl ester, 3-n-tert-butoxycarbonyl-n-methylamino propylamine, tert-butyl n-3-aminopropyl-n-methyl carbamate
2756543
tert-butyl N-(3-aminopropyl)-N-methylcarbamate
CC(C)(C)OC(=O)N(C)CCCN

It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color Yellow
Solubility Information Slightly soluble in water.
Formula Weight 188.27
Percent Purity 95%
Quantity 250 mg
Physical Form Liquid
Chemical Name or Material N-Boc-N-methyl-1,3-diaminopropane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'