missing translation for 'onlineSavingsMsg'
Learn More

n-Butylamine, 99+%, Thermo Scientific Chemicals

Catalog Number p-3735783
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal drum
109-73-9
C4H11N
73.13
MFCD00011690
HQABUPZFAYXKJW-UHFFFAOYSA-N
butylamine, n-butylamine, 1-butanamine, 1-aminobutane, 1-butylamine, monobutylamine, n-butylamin, mono-n-butylamine, norvalamine, 1-aminobutan
8007
CHEBI:43799
butan-1-amine
CCCCN
109-73-9
C4H11N
73.13
MFCD00011690
HQABUPZFAYXKJW-UHFFFAOYSA-N
butylamine, n-butylamine, 1-butanamine, 1-aminobutane, 1-butylamine, monobutylamine, n-butylamin, mono-n-butylamine, norvalamine, 1-aminobutan
8007
CHEBI:43799
butan-1-amine
CCCCN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point -49.0°C
Density 0.7400g/mL
Boiling Point 78.0°C
Flash Point −7°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3NH2
Refractive Index 1.4000 to 1.4020
Quantity 250 mL
Beilstein 04,156
Fieser 04,53
Merck Index 15,1545
Specific Gravity 0.74
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with alcohol and ether
Viscosity 0.5 mPa.s (20°C)
Formula Weight 73.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material n-Butylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Butylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'