missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzyloxy-L-methionine, 97%, Thermo Scientific™

Catalog Number p-4323697
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1152-62-1
C13H17NO4S
283.34
MFCD00065124
FPKHNNQXKZMOJJ-UHFFFAOYNA-N
z-met-oh, n-cbz-l-methionine, cbz-l-methionine, cbz-met-oh, z-l-methionine, n-carbobenzyloxy-l-methionine, n-carbobenzoxy-l-methionine, benzyloxycarbonyl-l-methionine, n-benzyloxy carbonyl-l-methionine, n-benzyloxycarbonyl-l-methionine
1550857
(2S)-4-methylsulfanyl-2-(phenylmethoxycarbonylamino)butanoic acid
CSCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
This item is not returnable. View return policy
1152-62-1
C13H17NO4S
283.34
MFCD00065124
FPKHNNQXKZMOJJ-UHFFFAOYNA-N
z-met-oh, n-cbz-l-methionine, cbz-l-methionine, cbz-met-oh, z-l-methionine, n-carbobenzyloxy-l-methionine, n-carbobenzoxy-l-methionine, benzyloxycarbonyl-l-methionine, n-benzyloxy carbonyl-l-methionine, n-benzyloxycarbonyl-l-methionine
1550857
(2S)-4-methylsulfanyl-2-(phenylmethoxycarbonylamino)butanoic acid
CSCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 67°C to 70°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Specific Rotation -27
Formula Weight 283.35
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material N-Carbobenzyloxy-L-methionine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'