missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzyloxy-L-methionine, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 67°C to 70°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (HPLC) |
Quantity | 5 g |
Specific Rotation | -27 |
Formula Weight | 283.35 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'