missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Cyclohexyl-2-nitrobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60086.03

Code : NEW

Additional Details : CAS Number : 77516-53-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416558

  • 448.12 € / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Cyclohexyl-2-nitrobenzenesulfonamide
77516-53-1
284.33
MFCD00436909

Specifications

N-Cyclohexyl-2-nitrobenzenesulfonamide
Solid
284.33
1g
77516-53-1
97%
MFCD00436909
Videos
SDS