missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Demethyllevomepromazine Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
N-Demethyllevomepromazine Hydrochloride, Mikromol
Specifications
Specifications
| Chemical Name or Material | N-Desmethyllevomepromazine Hydrochloride |
| CAS | 61733-92-4 |
| Analyte or Component Names | N-Desmethyllevomepromazine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H22 N2 O S . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | N-Demethyllevomepromazine Hydrochloride |
| InChI Formula | InChI=1S/C18H22N2OS.ClH/c1-13(11-19-2)12-20-15-6-4-5-7-17(15)22-18-9-8-14(21-3)10-16(18)20;/h4-10,13,19H,11-12H2,1-3H3;1H/t13-;/m1./s1 |
| Solution Type | Neat |
| SMILES | Cl.CNC[C@@H](C)CN1c2ccccc2Sc3ccc(OC)cc13 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction