missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmaleimide, 99+%, ACROS Organics™

Brand:  Acros Organics 156102500

Code : 29

Additional Details : CAS Number : 128-53-0 Weight : 0.30000kg Transport : UN number : 2928 Chem class : 6.1 Pack group : II

Quantity 250g

 View more versions of this product

Product Code. 11945581

  • 1394.00 € / 250g

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

128-53-0
125.13
n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide
CHEBI:44485
CCN1C(=O)C=CC1=O
C6H7NO2
HDFGOPSGAURCEO-UHFFFAOYSA-N
4362
1-ethylpyrrole-2,5-dione

Specifications

N-Ethylmaleimide
Authentic
210.0°C
41.0°C to 45.0°C
128-53-0
C6H7NO2
15,3877
Solubility in water: soluble. Other solubilities: 1115g/L toluene (20°C)
CCN1C(=O)C=CC1=O
125.13
CHEBI:44485
Crystalline Powder
73°C
Glass Bottle
White
250g
99% min. (GC)
21,IV,4629
n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide
HDFGOPSGAURCEO-UHFFFAOYSA-N
1-ethylpyrrole-2,5-dione
4362
125.13
99+%
Videos
SDS Documents
Certificates