missing translation for 'onlineSavingsMsg'
Learn More

n-Hexadecane, 99%, anhydrous, AcroSeal™, ACROS Organics™

Brand:  Acros Organics 436141000

Code : 29

Additional Details : CAS Number : 544-76-3 Weight : 0.13000kg

Packaging AcroSeal™ Glass Bottle
Quantity 100mL

 View more versions of this product

Product Code. 11964411

  • 84.99 € / 100mL

Estimated Shipment
05-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

544-76-3
226.44
n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
CHEBI:45296
CCCCCCCCCCCCCCCC
C16H34
DCAYPVUWAIABOU-UHFFFAOYSA-N
11006
hexadecane

Specifications

n-Hexadecane
135°C
AcroSeal™ Glass Bottle
0.773
0.005% max.
Colorless
100mL
98.5
98.5% min. (GC)
CH3(CH2)14CH3
Solubility in water: insoluble. Other solubilities: soluble in alcohol,acetone and ether
CCCCCCCCCCCCCCCC
226.44
CHEBI:45296
Liquid
0.7730g/mL
Authentic
1.4330 to 1.435
3.032 mPa.s (25°C)
287°C
18°C
544-76-3
100.0
C16H34
n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
DCAYPVUWAIABOU-UHFFFAOYSA-N
hexadecane
11006
226.44
99%
Videos
SDS Documents