missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, 97+%, for HPLC, Thermo Scientific Chemicals

Catalog Number 11934421
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
110-54-3
C6H14
86.18
MFCD02179311
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
8058
CHEBI:29021
hexane
CCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
110-54-3
C6H14
86.18
MFCD02179311
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
8058
CHEBI:29021
hexane
CCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -95.0°C
Density 0.6590g/mL
Boiling Point 69.0°C
Flash Point −22°C
Assay Percent Range 97% min. (GC)
Packaging Glass Bottle
Linear Formula CH3(CH2)4CH3
Refractive Index 1.3748 to 1.3810
Quantity 1 L
Merck Index 15,4732
Specific Gravity 0.659
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Viscosity 0.31 mPa.s (20°C)
Formula Weight 86.18
Percent Purity 97+%
Grade HPLC
Chemical Name or Material n-Hexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Hexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H361f-Suspected of damaging fertility.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P273-Avoid release to the environment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'