missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ n-Hexane, anhydrous, over molecular sieves, packaged under argon in resealable ChemSeal™ bottles

Brand:  Alfa Aesar™ 047104.K2

Code : NEW

Additional Details : CAS Number : 110-54-3 Weight : 1.00000kg Transport : UN number : 1208 Chem class : 3 Pack group : II

Quantity 1L

 View more versions of this product

Product Code. 15475065

  • 89.50 € / 1 L


Estimated Shipment date
from Supplier 20-06-2019
Add to basket

Description

Description

  • Packaged under Argon
Specifications

Chemical Identifiers

110-54-3
86.178
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD00009520
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC

Specifications

n-Hexane
110-54-3
CH3(CH2)4CH3
UN1208
14,4694
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18
0.659g/mL
1.3750
Colorless
1L
Anhydrous, over molecular sieves
C6H14
MFCD00009520
1730733
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CCCCCC
86.178
CHEBI:29021
Liquid
ChemSeal™ bottles
69°C
-95°C
Videos
SDS