Learn More
N'-Hydroxy-4-[(2-methyl-1,3-thiazol-4-yl)methoxy]benzenecarboximidamide, 95%, Maybridge™
Brand: Maybridge JM00487DA
Code : 31
Additional Details : CAS Number : 306935-19-3 Weight : 0.05100kg
Packaging | Amber Glass Bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
306935-19-3 | |
263.315 | |
QHRPKGPTUYZTBY-UHFFFAOYSA-N | |
9582192 | |
CC1=NC(=CS1)COC2=CC=C(C=C2)C(=NO)N |
C12H13N3O2S | |
MFCD00830779 | |
n'-hydroxy-4-2-methyl-1,3-thiazol-4-yl methoxy benzenecarboximidamide, 4-2-methylthiazol-4-ylmethoxy benzamidoxime, n'-hydroxy-4-2-methylthiazol-4-yl methoxy benzimidamide, hydroxyimino 4-2-methyl 1,3-thiazol-4-yl methoxy phenyl methylamine, z-n'-hydroxy-4-2-methyl-1,3-thiazol-4-yl methoxy benzene-1-carboximidamide, n'-hydroxy-4-2-methyl-1,3-thiazol-4-yl methoxy benzene-1-carboximidamide | |
N'-hydroxy-4-[(2-methyl-1,3-thiazol-4-yl)methoxy]benzenecarboximidamide |
Specifications
N'-Hydroxy-4-[(2-methyl-1, 3-thiazol-4-yl)methoxy]benzenecarboximidamide | |
1g | |
95.0 | |
C12H13N3O2S | |
n'-hydroxy-4-2-methyl-1,3-thiazol-4-yl methoxy benzenecarboximidamide, 4-2-methylthiazol-4-ylmethoxy benzamidoxime, n'-hydroxy-4-2-methylthiazol-4-yl methoxy benzimidamide, hydroxyimino 4-2-methyl 1,3-thiazol-4-yl methoxy phenyl methylamine, z-n'-hydroxy-4-2-methyl-1,3-thiazol-4-yl methoxy benzene-1-carboximidamide, n'-hydroxy-4-2-methyl-1,3-thiazol-4-yl methoxy benzene-1-carboximidamide | |
CC1=NC(=CS1)COC2=CC=C(C=C2)C(=NO)N | |
263.315 | |
263.32 |
Amber Glass Bottle | |
306935-19-3 | |
95% | |
MFCD00830779 | |
QHRPKGPTUYZTBY-UHFFFAOYSA-N | |
N'-hydroxy-4-[(2-methyl-1,3-thiazol-4-yl)methoxy]benzenecarboximidamide | |
9582192 | |
95% |