missing translation for 'onlineSavingsMsg'
Learn More

N-Lauroylsarcosine sodium salt, 95%, Thermo Scientific Chemicals

Catalog Number p-4228757
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Plastic Drum
137-16-6
C15H28NNaO3
293.38
MFCD00042728
KSAVQLQVUXSOCR-UHFFFAOYSA-M
sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
23668817
sodium;2-[dodecanoyl(methyl)amino]acetate
[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O
137-16-6
C15H28NNaO3
293.38
MFCD00042728
KSAVQLQVUXSOCR-UHFFFAOYSA-M
sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
23668817
sodium;2-[dodecanoyl(methyl)amino]acetate
[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94
CAS Max % 100
Melting Point 45.0°C to 47.0°C
Color White
pH 7.0 to 9.0
Loss on Drying 7% max. ()
Infrared Spectrum Authentic
Assay Percent Range 0.95
Packaging Glass bottle
Linear Formula NaCO2CH2N(CH3)CO(CH2)10CH3
Quantity 25 g
Merck Index 154401
Solubility Information (10% in water) Almost clear colorless to light yellow
Formula Weight 293.39
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Lauroylsarcosine sodium salt
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Lauroylsarcosine sodium salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'