missing translation for 'onlineSavingsMsg'
Learn More

N-Methoxy-N-methylbenzamide, 98%, Thermo Scientific™

Catalog Number p-4228775
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6919-61-5
C9H11NO2
165.19
UKERDACREYXSIV-UHFFFAOYSA-N
n-methyl-n-methoxybenzamide, benzamide, n-methoxy-n-methyl, n-methoxy-n-methyl-benzamide, methyl n-methylbenzohydroxamate, pubchem17496, acmc-1b5bp, n-methoxy-n-methylbenzamid, n-methoxy-n-methyl benzamide, ksc445c3n, n-methoxy-n-methylbenzamide
569575
CHEBI:59742
N-methoxy-N-methylbenzamide
CN(C(=O)C1=CC=CC=C1)OC
This item is not returnable. View return policy
6919-61-5
C9H11NO2
165.19
UKERDACREYXSIV-UHFFFAOYSA-N
n-methyl-n-methoxybenzamide, benzamide, n-methoxy-n-methyl, n-methoxy-n-methyl-benzamide, methyl n-methylbenzohydroxamate, pubchem17496, acmc-1b5bp, n-methoxy-n-methylbenzamid, n-methoxy-n-methyl benzamide, ksc445c3n, n-methoxy-n-methylbenzamide
569575
CHEBI:59742
N-methoxy-N-methylbenzamide
CN(C(=O)C1=CC=CC=C1)OC
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 85°C (1.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 165.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Methoxy-N-methylbenzamide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'