missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(4-nitrobenzyl)amine hydrochloride, 90%, Thermo Scientific™

Catalog Number p-3076415
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
19499-60-6
C8H11ClN2O2
202.638
MFCD06407995
LADGAFKIYVCZGN-UHFFFAOYSA-N
n-methyl-1-4-nitrophenyl methanamine hydrochloride, n-methyl-n-4-nitrobenzyl amine hydrochloride, n-methyl-4-nitrobenzylamine hydrochloride, methyl 4-nitrophenyl methyl amine hydrochloride, methyl 4-nitrophenyl methyl amine, chloride, n-methyl-n-4-nitrobenzyl amine hcl, n-methyl-1-4-nitrophenyl methanamine-hydrogen chloride 1/1
2794639
N-methyl-1-(4-nitrophenyl)methanamine;hydrochloride
CNCC1=CC=C(C=C1)[N+](=O)[O-].Cl
This item is not returnable. View return policy
19499-60-6
C8H11ClN2O2
202.638
MFCD06407995
LADGAFKIYVCZGN-UHFFFAOYSA-N
n-methyl-1-4-nitrophenyl methanamine hydrochloride, n-methyl-n-4-nitrobenzyl amine hydrochloride, n-methyl-4-nitrobenzylamine hydrochloride, methyl 4-nitrophenyl methyl amine hydrochloride, methyl 4-nitrophenyl methyl amine, chloride, n-methyl-n-4-nitrobenzyl amine hcl, n-methyl-1-4-nitrophenyl methanamine-hydrogen chloride 1/1
2794639
N-methyl-1-(4-nitrophenyl)methanamine;hydrochloride
CNCC1=CC=C(C=C1)[N+](=O)[O-].Cl
CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 202.64
Percent Purity 90%
Chemical Name or Material N-methyl-N-(4-nitrobenzyl)amine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'