missing translation for 'onlineSavingsMsg'
Learn More

N-Methyldiethanolamine, 99+%, Thermo Scientific Chemicals

Catalog Number 10553314
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
105-59-9
C5H13NO2
119.16
MFCD00002848
CRVGTESFCCXCTH-UHFFFAOYSA-N
n-methyldiethanolamine, n-methyliminodiethanol, methyldiethanolamine, n-methyldiethanolimine, n-methylaminodiglycol, 2,2'-methylimino diethanol, ethanol, 2,2'-methylimino bis, methyl diethanolamine, n-methyl-2,2'-iminodiethanol, 2,2'-methyliminodiethanol
7767
CN(CCO)CCO
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
105-59-9
C5H13NO2
119.16
MFCD00002848
CRVGTESFCCXCTH-UHFFFAOYSA-N
n-methyldiethanolamine, n-methyliminodiethanol, methyldiethanolamine, n-methyldiethanolimine, n-methylaminodiglycol, 2,2'-methylimino diethanol, ethanol, 2,2'-methylimino bis, methyl diethanolamine, n-methyl-2,2'-iminodiethanol, 2,2'-methyliminodiethanol
7767
CN(CCO)CCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point -21°C
Color Colorless to Yellow
Density 1.0380 g/mL
Boiling Point 243°C
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (HOCH2CH2)2NCH3
Refractive Index 1.4675 to 1.4695
Quantity 2.5 kg
Beilstein 04, 284
Specific Gravity 1.038
Solubility Information Solubility aq. soln.: miscible. Other solubilities: miscible with benzene,soluble in most organic solvents (alcohol,esters,aceton and chlorinated hydrocarbons),soluble in diethylether and aliphatic hydrocarbons
IUPAC Name 2-[(2-hydroxyethyl)(methyl)amino]ethan-1-ol
Viscosity 101 mPa.s (20°C)
Formula Weight 119.16
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material N-Methyldiethanolamine, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • N-Methyldiethanolamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'