missing translation for 'onlineSavingsMsg'
Learn More

N-Methylindole-3-boronic acid pinacol ester, 97%, ACROS Organics™

Brand:  Acros Organics 441010010

Code : 29

Additional Details : CAS Number : 683229-61-0 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 11956711

  • 170.00 € / 1g

Estimated Shipment
23-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

683229-61-0
257.14
1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-indole, n-methylindole-3-boronic acid pinacol ester, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indole, n-methylindole-3-boronic acid, pinacol ester, 1-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 1-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborole-2-yl-1h-indole, 1h-indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indole
C15H20BNO2
MCCCOWZSGBKRCG-UHFFFAOYSA-N
23058003
B1(OC(C(O1)(C)C)(C)C)C2=CN(C3=CC=CC=C23)C

Specifications

N-Methylindole-3-boronic acid pinacol ester
Glass bottle
106°C to 112°C
683229-61-0
100.0
C15H20BNO2
MCCCOWZSGBKRCG-UHFFFAOYSA-N
1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indole
23058003
Low Melting Solid
Authentic
Brown to White
1g
96.0
96% min. (GC)
1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-indole, n-methylindole-3-boronic acid pinacol ester, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indole, n-methylindole-3-boronic acid, pinacol ester, 1-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indole, 1-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborole-2-yl-1h-indole, 1h-indole, 1-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
B1(OC(C(O1)(C)C)(C)C)C2=CN(C3=CC=CC=C23)C
257.14
257.14
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • N-Methylindole-3-boronic acid pinacol ester

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS Documents