Learn More
N,N'-Bis(2-hydroxyethyl)ethylenediamine, 95%, ACROS Organics™
Brand: Acros Organics 345180100
Code : 29
Additional Details : CAS Number : 4439-20-7 Weight : 0.06000kg Transport : UN number : 3259 Chem class : 8 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 10g |
Chemical Identifiers
4439-20-7 | |
148.2 | |
GFIWSSUBVYLTRF-UHFFFAOYSA-N | |
78179 | |
C(CNCCO)NCCO |
C6H16N2O2 | |
MFCD00002842 | |
n,n'-bis 2-hydroxyethyl ethylenediamine, n,n'-diethanolethylenediamine, ethanol, 2,2'-1,2-ethanediyldiimino bis, 1,2-bis 2-hydroxyethyl amino ethane, 3,6-diazaoctane-1,8-diol, 2,2'-ethane-1,2-diylbis azanediyl diethanol, n,n'-bis 2-hydroxyethyl ethylene diamine, n,n'-bis 2-hydroxyethyl-ethylenediamine, 2-2-2-hydroxyethylamino ethylamino ethanol | |
2-[2-(2-hydroxyethylamino)ethylamino]ethanol |
Specifications
N, N'-Bis(2-hydroxyethyl)ethylenediamine | |
Glass bottle | |
92°C to 99°C | |
4439-20-7 | |
100.0 | |
C6H16N2O2 | |
MFCD00002842 | |
GFIWSSUBVYLTRF-UHFFFAOYSA-N | |
2-[2-(2-hydroxyethylamino)ethylamino]ethanol | |
78179 | |
Powder and/or Chunks |
Authentic | |
White to Yellow | |
10g | |
94.0 | |
95% | |
(HOCH2CH2NHCH2-)2 | |
n,n'-bis 2-hydroxyethyl ethylenediamine, n,n'-diethanolethylenediamine, ethanol, 2,2'-1,2-ethanediyldiimino bis, 1,2-bis 2-hydroxyethyl amino ethane, 3,6-diazaoctane-1,8-diol, 2,2'-ethane-1,2-diylbis azanediyl diethanol, n,n'-bis 2-hydroxyethyl ethylene diamine, n,n'-bis 2-hydroxyethyl-ethylenediamine, 2-2-2-hydroxyethylamino ethylamino ethanol | |
C(CNCCO)NCCO | |
148.2 | |
148.2 | |
95% |
Safety and Handling
- N,N'-Bis(2-hydroxyethyl)ethylenediamine
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.