missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diethyl-p-phenylenediamine oxalate, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 95.0 |
CAS Max % | 100.0 |
Melting Point | 145.0°C to 150.0°C |
Color | Brown to Gray or Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% |
Packaging | Glass bottle |
Quantity | 10 g |
Formula Weight | 209.27 |
Percent Purity | 96% |
Show More |
Safety and Handling
Product Identifier
- N,N-Diethyl-p-phenylenediamine oxalate
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'