Learn More
N,N-Diisopropylcarbamoyl chloride, 98%, ACROS Organics™
Brand: Acros Organics 314520250
Code : 29
Additional Details : CAS Number : 19009-39-3 Weight : 0.07500kg Transport : UN number : 3261 Chem class : 8 Pack group : II
Packaging | Glass bottle |
---|---|
Quantity | 25g |
Chemical Identifiers
19009-39-3 | |
163.65 | |
diisopropylcarbamoyl chloride, n,n-diisopropylcarbamoyl chloride, diisopropylcarbamic chloride, diisopropylcarbamyl chloride, n,n-di propan-2-yl carbamoyl chloride, acmc-20ak8x, diisopropyl carbamyl chloride, di-i-propylcarbamoyl chloride, n,n-diisopropylchloroformamide, n,n-diisopropylcarbamoylchloride | |
N,N-di(propan-2-yl)carbamoyl chloride |
C7H14ClNO | |
RSAFAYLZKCYUQW-UHFFFAOYSA-N | |
87890 | |
CC(C)N(C(C)C)C(=O)Cl |
Specifications
N, N-Diisopropylcarbamoyl chloride | |
Glass bottle | |
Colorless to Yellow or White to Yellow | |
25g | |
97.5 | |
98% | |
[(CH3)2CH]2NCOCl | |
RSAFAYLZKCYUQW-UHFFFAOYSA-N | |
N,N-di(propan-2-yl)carbamoyl chloride | |
87890 | |
Crystalline Powder or To Low Melting Solid |
Authentic | |
90.0°C to 93.0°C (15.0mmHg) | |
58.1°C to 59.1°C | |
19009-39-3 | |
100.0 | |
C7H14ClNO | |
diisopropylcarbamoyl chloride, n,n-diisopropylcarbamoyl chloride, diisopropylcarbamic chloride, diisopropylcarbamyl chloride, n,n-di propan-2-yl carbamoyl chloride, acmc-20ak8x, diisopropyl carbamyl chloride, di-i-propylcarbamoyl chloride, n,n-diisopropylchloroformamide, n,n-diisopropylcarbamoylchloride | |
CC(C)N(C(C)C)C(=O)Cl | |
163.65 | |
163.65 | |
98% |
Safety and Handling
- N,N-Diisopropylcarbamoyl chloride, 98%
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.