missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-3523507
21.15 EUR valid until 2024-03-31
Use promo code "22193" to get your promotional price.


Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
68-12-2
C3H7NO
73.10
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
68-12-2
C3H7NO
73.10
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O

N,N-Dimethylformamide, 99.8%, C3H7NO, CAS Number-68-12-2, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dwumetyloformamid, dimethylformamide, dmfa, dimethyl formamide, dimetilformamide, dmf, dimethylformamid, 1L, 0.01 % max., Vapor Pressure: 4.9mbar at 20 deg.C, 04, 58

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -61.0°C
CAS Min % 99.75
Color White to Beige
CAS Max % 100.0
Assay Percent Range 99.8%
Linear Formula HCON(CH3)2
Beilstein 04, 58
Fieser 01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144
Merck Index 15, 3267
Solubility Information Solubility in water: soluble. Other solubilities: miscible with most organic solvents
Formula Weight 73.09
Percent Purity 99.8%
Grade Extra Dry over Molecular Sieve
Decomposition Information >350°C
Density 0.9450g/mL
Boiling Point 153.0°C
Quantity 100 mL
Flash Point 58°C
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Physical Form Crystalline Powder
Refractive Index 1.4290 to 1.4320
Specific Gravity 0.945
Vapor Pressure Vapor Pressure: 4.9mbar at 20°C
Viscosity 0.8 mPa.s (20°C)
Water 0.005% Max. (Coulometric)
Chemical Name or Material N,N-Dimethylformamide, water <50 ppm
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'