missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylhexylamine, 99%, Thermo Scientific Chemicals

Catalog Number p-3757519
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4385-04-0
C8H19N
129.25
MFCD00009522
QMHNQZGXPNCMCO-UHFFFAOYSA-N
n,n-dimethylhexylamine, hexyldimethylamine, 1-hexanamine, n,n-dimethyl, 1-dimethylaminohexane, n,n-dimethyl-n-hexylamine, dimethylhexylamine, hexyl-dimethyl-amine, n-hexyl dimethylamine, hexanamine, n,n-dimethyl, acmc-1ct1t
78090
N,N-dimethylhexan-1-amine
CCCCCCN(C)C
This item is not returnable. View return policy
4385-04-0
C8H19N
129.25
MFCD00009522
QMHNQZGXPNCMCO-UHFFFAOYSA-N
n,n-dimethylhexylamine, hexyldimethylamine, 1-hexanamine, n,n-dimethyl, 1-dimethylaminohexane, n,n-dimethyl-n-hexylamine, dimethylhexylamine, hexyl-dimethyl-amine, n-hexyl dimethylamine, hexanamine, n,n-dimethyl, acmc-1ct1t
78090
N,N-dimethylhexan-1-amine
CCCCCCN(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.7440g/mL
Boiling Point 146°C to 150°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)5N(CH3)2
Refractive Index 1.4126 to 1.4146
Quantity 5 g
Specific Gravity 0.744
Formula Weight 129.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylhexylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'