missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenylurea, 97%, Thermo Scientific Chemicals

Catalog Number p-4323686
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
102-07-8
C13H12N2O
212.25
MFCD00003017
GWEHVDNNLFDJLR-UHFFFAOYSA-N
n,n'-diphenylurea, carbanilide, diphenylurea, diphenylcarbamide, s-diphenylurea, urea, n,n'-diphenyl, sym-diphenylurea, acardite i, acardite, n-phenyl-n'-phenylurea
7595
CHEBI:41320
1,3-diphenylurea
O=C(NC1=CC=CC=C1)NC1=CC=CC=C1
This item is not returnable. View return policy
102-07-8
C13H12N2O
212.25
MFCD00003017
GWEHVDNNLFDJLR-UHFFFAOYSA-N
n,n'-diphenylurea, carbanilide, diphenylurea, diphenylcarbamide, s-diphenylurea, urea, n,n'-diphenyl, sym-diphenylurea, acardite i, acardite, n-phenyl-n'-phenylurea
7595
CHEBI:41320
1,3-diphenylurea
O=C(NC1=CC=CC=C1)NC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 232.0°C to 238.0°C
Color White
Boiling Point 260.0°C to 262.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5NHCONHC6H5
Quantity 25 g
Beilstein 12,352
Merck Index 15,1786
Formula Weight 212.25
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N, N'-Diphenylurea
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'