missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium hexafluorophosphate, 98%, Thermo Scientific™

Catalog Number p-4229335
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
265651-18-1
C9H16F6N3O3P
359.21
MFCD01863753
STWZCCVNXFLDDD-UHFFFAOYSA-N
hstu, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium hexafluorophosphate, hstu n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium hexafluorophosphate, ambotzrl-1039, o-n-succinimidyl-1,1,3,3-tetramethyluronium hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronim hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uroniumhexafluorophosphate
16211151
F[P-](F)(F)(F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C
This item is not returnable. View return policy
265651-18-1
C9H16F6N3O3P
359.21
MFCD01863753
STWZCCVNXFLDDD-UHFFFAOYSA-N
hstu, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium hexafluorophosphate, hstu n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium hexafluorophosphate, ambotzrl-1039, o-n-succinimidyl-1,1,3,3-tetramethyluronium hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronim hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uroniumhexafluorophosphate
16211151
F[P-](F)(F)(F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C
Coupling reagent
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 213°C to 215°C
Color White
Elemental Analysis 29.34 to 30.85% C, 11.41 to 11.99% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 25 g
IUPAC Name [(dimethylamino)[(2,5-dioxopyrrolidin-1-yl)oxy]methylidene]dimethylazanium; hexafluoro-λ⁵-phosphanuide
Formula Weight 359.21
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium hexafluorophosphate
Show More Show Less
compliance-icons
Product Identifier
  • HSTU
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'