missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitrosodimethylamine (NDMA) 5000 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | N-Nitrosodimethylamine |
| CAS | 62-75-9 |
| Analyte or Component Names | N-Nitrosodimethylamine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C2 H6 N2 O |
| Concentration | 5000 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Methanamine, N-methyl-N-nitroso-, Dimethylamine, N-nitroso- (8CI), N-Methyl-N-nitrosomethanamine, DMN, DMNA, Dimethylnitrosamine, N-Nitroso-N,N-dimethylamine, N-Nitrosodimethylamine, NDMA, NSC 23226, N-Nitroso-dimethylamine |
| InChI Formula | InChI=1S/C2H6N2O/c1-4(2)3-5/h1-2H3 |
| SMILES | CN(C)N=O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction