missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Silylation reagent
Specifications
Color | Colorless to Yellow |
Density | 0.9740g/mL |
Boiling Point | 145.0°C to 147.0°C |
Flash Point | 34°C |
Linear Formula | CF3C[=NSi(CH3)3]OSi(CH3)3 |
Quantity | 500 g |
Specific Gravity | 0.974 |
Solubility Information | Solubility in water: hydrolysis |
Formula Weight | 257.4 |
Percent Purity | ≥98.0% (excluding TMCS), 1.5% max. TMCS |
Show More |
Safety and Handling
Product Identifier
- N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H314-Causes severe skin burns and eye damage.
- H319-Causes serious eye irritation.
- H226-Flammable liquid and vapour.
- EUH014-Reacts violently with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'