missing translation for 'onlineSavingsMsg'
Learn More

N-Propyl Acetate, Extra Pure, SLR, Fisher Chemical™

Product Code. p-7073463 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10396140

Brand: Fisher Chemical P/7600/08

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 109-60-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00009372
InChI Key YKYONYBAUNKHLG-UHFFFAOYSA-N
Synonym n-propyl acetate, acetic acid, propyl ester, propyl ethanoate, 1-acetoxypropane, n-propyl ethanoate, 1-propyl acetate, acetic acid n-propyl ester, octan propylu, propylacetate, acetic acid propyl ester
PubChem CID 7997
ChEBI CHEBI:40116
IUPAC Name propyl acetate
SMILES CCCOC(C)=O

Specifications

Melting Point -95°C
Color Colorless
Boiling Point 102°C
Packaging Amber glass bottle
Quantity 500 mL
Vapor Density 3.5
Formula Weight 102.13g/mol
Vapor Pressure 33mbar at 20°C
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • n-Propyl acetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.