missing translation for 'onlineSavingsMsg'
Learn More

n-Propyltrichlorosilane, 98%, ACROS Organics™

Brand:  Acros Organics 147411000

Code : 29

Additional Details : CAS Number : 141-57-1 Weight : 0.16900kg Transport : UN number : 1816 Chem class : 8 Pack group : II


Packaging Glass bottle
Quantity 100mL

 View more versions of this product

Product Code. 11955801

  • 29.03 € / 100mL

Estimated Shipment
27-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

141-57-1
177.53
propyltrichlorosilane, trichloro propyl silane, n-propyltrichlorosilane, silane, trichloropropyl, propyl trichlorosilane, trichloro-n-propylsilane, silane, propyltrichloro, unii-x16g85fi4y, hsdb 889, silane,trichloropropyl
trichloro(propyl)silane
C3H7Cl3Si
DOEHJNBEOVLHGL-UHFFFAOYSA-N
8850
CCC[Si](Cl)(Cl)Cl

Specifications

n-Propyltrichlorosilane
31°C
Glass bottle
1.19
Colorless to Brown
141-57-1
100.0
C3H7Cl3Si
04,36
Solubility in water: reacts
CCC[Si](Cl)(Cl)Cl
177.53
177.53
98%
1.1900g/mL
Authentic
1.195
123.0°C to 124.0°C
100mL
97.5
97.5% min. (GC)
CH3CH2CH2SiCl3
propyltrichlorosilane, trichloro propyl silane, n-propyltrichlorosilane, silane, trichloropropyl, propyl trichlorosilane, trichloro-n-propylsilane, silane, propyltrichloro, unii-x16g85fi4y, hsdb 889, silane,trichloropropyl
DOEHJNBEOVLHGL-UHFFFAOYSA-N
trichloro(propyl)silane
8850
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • n-Propyltrichlorosilane

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents