missing translation for 'onlineSavingsMsg'
Learn More

N-Vinyl-2-pyrrolidone, 99%, stabilized with NaOH, Acros Organics

Brand:  Acros Organics 140920100

Code : NEW

Additional Details : CAS Number : 88-12-0 Weight : 10.43000kg


Quantity 10L

 View more versions of this product

Product Code. 15264634

  • 1171.00 € / 10 L

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!



Specifications

Chemical Identifiers

88-12-0
111.14
WHNWPMSKXPGLAX-UHFFFAOYSA-N
6917
1-ethenylpyrrolidin-2-one
C6H9NO
MFCD00003197
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
CHEBI:82551
C=CN1CCCC1=O

Specifications

N-Vinyl-2-pyrrolidone
95°C
1.5119 to 1.5139
0.1% flaked caustic soda
0.2% max. (K.F.)
Colorless to Yellow
10L
88-12-0
100.0
C6H9NO
15,7814
Solubility in water: 52.1g/L (25°C). Other solubilities: miscible with acetone,diethyl ether,ethanol,,toluene,benzene,very good soluble in organic solvents
C=CN1CCCC1=O
111.14
CHEBI:82551
Liquid
1.0430g/mL
Authentic
1.043
2.4 mPa.s (20°C)
218°C
13°C to 14°C
98%
98.5
0.2% max. (Pyrrolidone) (GC)
MFCD00003197
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
WHNWPMSKXPGLAX-UHFFFAOYSA-N
1-ethenylpyrrolidin-2-one
6917
111.14
99%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • N-Vinyl-2-pyrrolidone, stabilized

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Documents