missing translation for 'onlineSavingsMsg'
Learn More

N1-(4-Amino-2-methylphenyl)acetamide, 90%, Maybridge™

Brand:  Maybridge BTB06485EA

Code : 31

Additional Details : CAS Number : 56891-59-9 Weight : 0.06000kg

Packaging Amber Glass Bottle
Quantity 10g

 View more versions of this product

Product Code. 10028901

  • 185.73 € / 10g

Estimated Shipment
15-06-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

56891-59-9
164.208
GWFPMSIIVJMYRZ-UHFFFAOYSA-N
314338
CC1=C(C=CC(=C1)N)NC(=O)C
C9H12N2O
MFCD00276633
n-4-amino-2-methylphenyl acetamide, n1-4-amino-2-methylphenyl acetamide, 4'-amino-2'-methylacetanilide, n-4-amino-2-methyl-phenyl-acetamide, 4-acetamido-m-toluidine, 4-acetylamino-m-toluidine, 4-acetamido-3-methylaniline, 4-acetamino-3-methylaniline, acetamide, n-4-amino-2-methylphenyl
N-(4-amino-2-methylphenyl)acetamide

Specifications

N1-(4-amino-2-methylphenyl)acetamide
10g
89.5
90%
MFCD00276633
GWFPMSIIVJMYRZ-UHFFFAOYSA-N
N-(4-amino-2-methylphenyl)acetamide
314338
90%
Amber Glass Bottle
56891-59-9
100.0
C9H12N2O
n-4-amino-2-methylphenyl acetamide, n1-4-amino-2-methylphenyl acetamide, 4'-amino-2'-methylacetanilide, n-4-amino-2-methyl-phenyl-acetamide, 4-acetamido-m-toluidine, 4-acetylamino-m-toluidine, 4-acetamido-3-methylaniline, 4-acetamino-3-methylaniline, acetamide, n-4-amino-2-methylphenyl
CC1=C(C=CC(=C1)N)NC(=O)C
164.208
164.21
Videos
SDS Documents