missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ 870 PFA Tubing

Utilize translucent 870 tubing for transferring aggressive and corrosive chemical fluids. Useful over a very wide temperature range (-270° to +260°C) with low water absorption.

Brand:  Thermo Scientific™ 8051-0250

Code : NEW

Additional Details : Weight : 0.96000kg

 View more versions of this product

Product Code. 11314695

  • 627.00 € / 15m
In Stock
to see stock.
Add to basket

Description

Description

Utilize translucent Thermo Scientific™ Nalgene™ 870 Tubing for transferring aggressive and corrosive chemical fluids. Useful over a very wide temperature range (-270° to +260°C) with low water absorption. Excellent for low-pressure use with mechanical, compression fittings at room tempearature. Food-grade materials comply with 21 CFR 177.1520 for food packaging.

  • Wider temperature range -270° to +260°C (-518° to +500°F)
  • High level of inertness for high purity applications
  • Sterilization by autoclaving, gas, dry heat and disinfectants but not by radiations
  • Two 25-ft. coils per case except larger sizes
  • Autoclavable/PFA

Note:

Do not sterilize by radiation. Should not be used with molten alkali metals, elemental fluorine and fluorine precursors at elevated temperatures or concentrated perchloric acid.

Specifications

Specifications

Semi-Rigid Tubing
Autoclavable
1.2 mm
1/4 in.
50 ft.
3/64 in.
25
Durometer 60(D)
290 psig at 73°F (23°C), 145psig at 185°F (85°C)
260°C
Food-Grade materials comply with 21 CFR 177.1520 for food packaging
3/64 in.
PFA
6.4 mm
15.24 m
0.12 cm
Transparent
Bend Radius 10xOD
500°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers