missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Carboy Replacement Screw Closures and Gaskets

Catalog Number p-4521535
30.10 EUR valid until 2024-03-30
Use promo code "21648" to get your promotional price.


Click to view available options
Description:
Replacement Closure
Replacement Closure w/Strap
Sealing TPE (thermoplastic elastomer) Gasket
Material:
HDPE
Polypropylene
Silicone Grommet
TPE
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price

Secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys provide leakproof† service

  • Made to fit Nalgene Carboys
TRUSTED_SUSTAINABILITY
Closure Material Polypropylene
Closure Size 100 mm
For Use With (Equipment) Nalgene narrow-mouth and wider-mouth bottles
Product Type Closure
Color White
Description Replacement Closure
Material Polypropylene
No. per Case 12
Closure Included Yes
Mouth Narrow, Wide
Shape Round
Show More Show Less

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'