missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Centrifuge Tubes Made of Teflon™ FEP with ETFE Closure

Conduct high-speed centrifugation of aggressive chemicals with these tubes, suited for chloroform and phenol extractions in refrigerated machines up to 50,000 x g.

Brand:  Thermo Scientific™ 3114-0030

Code : 55

Additional Details : Weight : 0.52200kg

 View more versions of this product

Product Code. 10018700

Quantity Price
1 109.48 € / Pack of 2
10 99.38 € / Pack of 2
In stock, ready to ship: 7

Add to basket

Alert:

Note: Must be filled to 100% of total capacity for centrifugation


Description

Description

Conduct high-speed centrifugation of aggressive chemicals with Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made of Teflon™ FEP with ETFE Closure, specifically suited for chloroform and phenol extractions.

Features:

  • Teflon™ FEP, Tefzel™ ETFE screw closure
  • Withstand temperatures from -100° to +150°C (-148° to +302°F)
  • Leakproof† Tefzel ETFE closures prevent material loss under ordinary use for most spins <10,000 × g
  • Sealing caps available for 30mL and 50mL size to assure leakproof service especially at high speeds >10,000 × g or when spinning hazardous materials
  • Autoclavable/biohazard/leakproof†/PTFE FEP

Ordering Information:

Optional sealing caps available: DS3131-0020 (for 30mL tube) and DS3131-0024 (for 50mL tube). Cat. No. 3114-0050 has a brim capacity of only 42mL but is designed to fit standard rotor.

Includes:

Tefzel ETFE screw caps. Additional sealing cap assembly available separately (DS3131-series).

Specifications

Specifications

25.7 mm
Screw Cap
93.0mm
30 mL
10
Non-sterile
No
Centrifuge Tube
Round
No
ETFE
Teflon FEP
2
Autoclavable
Round
50,000 x g with sealing cap assembly
Videos
SDS
Documents
Certificates
View accessory products
Special Offers

Special Offers

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.