missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes w/Sealing Cap

View contents easily using these strong, transparent polycarbonate centrifuge tubes, available with sealing caps for use up to 50,000 x g.

Brand:  Thermo Scientific™ 3138-0016

103.64 EUR valid until 2024-04-26
Use promo code "21699" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.13500kg

 View more versions of this product

Product Code. 10179900

  • 173.40 € / Pack of 10
Estimated Shipment: 19-11-2024
to see stock.
Add to basket

Alert:

Autoclaving repeatedly will shorten the life of polycarbonate tubes. Tubes must be filled to at least 80% capacity for proper performance.


Description

Description

Easily view contents using strong, transparent Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes, equipped with sealing caps for leakproof† performance up to 50,000 x g.

Features:
  • Withstand temperatures from -135° to +135°C (-211° to +275°F)
  • Round bottom to fit high speed rotors
  • Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges
  • Come with PP sealing caps with silicone gaskets for extra leak protection when spinning >10,000 x g or spinning hazardous materials
  • Autoclavable; transparent

Ordering Information:

Sealing caps also sold separately for 30 and 50mL sizes.

Specifications

Specifications

Polypropylene, red rubber
Polycarbonate
10
Autoclavable
107.2 mm
Screw Cap
18.2 mm
Centrifuge Tube
16 mL
50
Non-sterile
Round
Round
No
No
50,000 x g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers