missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dimethoxybenzene, 99%, Thermo Scientific Chemicals
31.90 € - 113.00 €
Chemical Identifiers
CAS | 151-10-0 |
---|---|
Molecular Formula | C8H10O2 |
Molecular Weight (g/mol) | 138.17 |
MDL Number | MFCD00008384 |
InChI Key | DPZNOMCNRMUKPS-UHFFFAOYSA-N |
Synonym | m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu |
PubChem CID | 9025 |
IUPAC Name | 1,3-dimethoxybenzene |
SMILES | COC1=CC(OC)=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10784451
|
Thermo Scientific Acros
155231000 |
100 g | Glass bottle |
31.90 €
100g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
View alternative products » | |||||||||
10644632
|
Thermo Scientific Acros
155235000 |
500 g | Glass bottle |
113.00 €
500g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
View alternative products » | |||||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
151-10-0 | |
138.17 | |
DPZNOMCNRMUKPS-UHFFFAOYSA-N | |
9025 | |
COC1=CC(OC)=CC=C1 |
C8H10O2 | |
MFCD00008384 | |
m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu | |
1,3-dimethoxybenzene |
Specifications
1, 3-Dimethoxybenzene | |
151-10-0 | |
100.0 | |
1.0500g/mL | |
87°C | |
98.5% min. (GC) | |
C8H10O2 | |
C6H4(OCH3)2 | |
100 g | |
1.05 | |
Solubility in water: immiscible | |
COC1=CC(OC)=CC=C1 | |
138.17 | |
138.17 | |
Liquid |
99% | |
98.5 | |
Colorless to Brown | |
85.0°C to 87.0°C (7.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5230 to 1.5250 | |
MFCD00008384 | |
06, 813 | |
m-dimethoxybenzene, resorcinol dimethyl ether, benzene, 1,3-dimethoxy, 3-methoxyanisole, benzene, m-dimethoxy, dimethyl resorcinol, m-methoxyanisole, dimethylresorcinol, methoxyanisole, m, dimethylether resorcinolu | |
DPZNOMCNRMUKPS-UHFFFAOYSA-N | |
1,3-dimethoxybenzene | |
9025 | |
99% |
RUO – Research Use Only