missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Polycarbonate, Validation Bottle
Click to view available options
Capacity (Metric):
30 mL
250 mL
2 L
Unit Size:
Pack of 12
Pack of 30
Pack of 6
Description
Perform QA lot sample storage of high-valued pharmaceuticals using Thermo Scientific™ Nalgene™ Polycarbonate Validation Bottles. Small-volume bottles are molded of the same materials as larger carboys, but require a lower volume of expensive reagents to perform validation testing.
Autoclavable, transparent polycarbonate construction allows for convenient material compatibility validation.
Includes:
White polypropylene screw closure and TPE gasket
Specifications
Specifications
| Autoclavable | Autoclavable |
| Capacity (English) | 1 oz. |
| Capacity (Metric) | 30 mL |
| Closure Material | Polypropylene |
| Closure Size | 20-415 |
| Color | White (Translucent) |
| Diameter (Metric) Inner Neck | 14 mm |
| Diameter (Metric) Outer | 32 mm |
| For Use With (Application) | Perform validation testing |
| Height (Metric) With Closure | 75 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction